missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-picoline, 98%

Product Code. 10222722 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
This item is not returnable. View return policy

Product Code. 10222722

Brand: Thermo Scientific Acros 368840050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3678-62-4 | C6H6ClN | 127.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3678-62-4
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00023418
InChI Key MZVSTDHRRYQFGI-UHFFFAOYSA-N
Synonym 2-chloro-4-picoline, pyridine, 2-chloro-4-methyl, 2-chloro-4-methyl-pyridine, 2-chloro-4-methyl pyridine, 2-chloro-gamma-picoline, 4-picoline, 2-chloro, zlchem 438, pubchem1192, acmc-209iop
PubChem CID 77248
IUPAC Name 2-chloro-4-methylpyridine
SMILES CC1=CC(=NC=C1)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1510g/mL
Boiling Point 194°C to 195°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5280 to 1.5300 (20°C, 589nm)
Quantity 5 mL
Beilstein 20,241
Specific Gravity 1.151
Formula Weight 127.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-picoline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-4-picoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.