missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrobenzonitrile, 97%, Thermo Scientific™

Product Code. 10021894
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10021894 25 g Amber Glass Bottle 25g
10415413 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10021894 Supplier Thermo Scientific Maybridge Supplier No. XAX00005EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 28163-00-0
Molecular Formula C7H3ClN2O2
Molecular Weight (g/mol) 182.56
MDL Number MFCD00052203
InChI Key ZIGQFRQZYVQQDR-UHFFFAOYSA-N
Synonym 2-chloro-4-nitro-benzonitrile, benzonitrile, 2-chloro-4-nitro, 2-chloro-4-nitrobenzenecarbonitrile, pubchem18759, acmc-1ceg9, 2-chloro-4-nitrobenzonitril, chloronitrobenzenecarbonitrile, 3-chloro-4-cyanonitrobenzene, ksc497k1r
PubChem CID 141401
IUPAC Name 2-chloro-4-nitrobenzonitrile
SMILES [O-][N+](=O)C1=CC=C(C#N)C(Cl)=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 182.57
Percent Purity 97%
Chemical Name or Material 2-chloro-4-nitrobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.