missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylpyridine, 98%, Thermo Scientific™

Product Code. 15427088
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15427088 5 mL 5mL
15437088 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427088 Supplier Thermo Scientific Alfa Aesar Supplier No. H60607.AA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3678-62-4
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.571
MDL Number MFCD00023418
InChI Key MZVSTDHRRYQFGI-UHFFFAOYSA-N
Synonym 2-chloro-4-picoline, pyridine, 2-chloro-4-methyl, 2-chloro-4-methyl-pyridine, 2-chloro-4-methyl pyridine, 2-chloro-gamma-picoline, 4-picoline, 2-chloro, zlchem 438, pubchem1192, acmc-209iop
PubChem CID 77248
IUPAC Name 2-chloro-4-methylpyridine
SMILES CC1=CC(=NC=C1)Cl

Specifications

Density 1.142
Boiling Point 194°C to 195°C
Refractive Index 1.529
Quantity 5 mL
Formula Weight 127.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.