missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(methylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number 15422286
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1451391-89-1
C8H9BClNO3
213.424
MFCD22125174
LSMHGCJWZPHACP-UHFFFAOYSA-N
2-chloro-4-methylcarbamoyl phenylboronic acid, 2-chloro-4-methylcarbamoyl benzeneboronic acid, 2-chloro-4-methylcarbamoyl phenyl boronic acid
73996177
[2-chloro-4-(methylcarbamoyl)phenyl]boronic acid
B(C1=C(C=C(C=C1)C(=O)NC)Cl)(O)O
This item is not returnable. View return policy
1451391-89-1
C8H9BClNO3
213.424
MFCD22125174
LSMHGCJWZPHACP-UHFFFAOYSA-N
2-chloro-4-methylcarbamoyl phenylboronic acid, 2-chloro-4-methylcarbamoyl benzeneboronic acid, 2-chloro-4-methylcarbamoyl phenyl boronic acid
73996177
[2-chloro-4-(methylcarbamoyl)phenyl]boronic acid
B(C1=C(C=C(C=C1)C(=O)NC)Cl)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 213.43
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(methylcarbamoyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'