Learn More
2-chloro-4-methylaniline, 98%, Thermo Scientific Chemicals
£21.50 - £206.77
Chemical Identifiers
CAS | 615-65-6 |
---|---|
Molecular Formula | C7H8ClN |
Molecular Weight (g/mol) | 141.6 |
MDL Number | MFCD00007666 |
InChI Key | XGYLSRFSXKAYCR-UHFFFAOYSA-N |
Synonym | 2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene |
PubChem CID | 12007 |
IUPAC Name | 2-chloro-4-methylaniline |
SMILES | CC1=CC(=C(C=C1)N)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10427392
|
Thermo Scientific Acros
154670050 |
5 mL | Glass bottle |
£21.50
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10113400
|
Thermo Scientific Acros
154670250 |
25 mL | Glass bottle |
£65.60
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10289503
|
Thermo Scientific Acros
154671000 |
100 mL | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
615-65-6 | |
141.6 | |
XGYLSRFSXKAYCR-UHFFFAOYSA-N | |
12007 | |
CC1=CC(=C(C=C1)N)Cl |
C7H8ClN | |
MFCD00007666 | |
2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene | |
2-chloro-4-methylaniline |
Specifications
615-65-6 | |
100.0 | |
Brown to Yellow | |
223°C to 225°C | |
Authentic | |
C7H8ClN | |
1.5740 to 1.576 | |
12, 989 | |
2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene | |
CC1=CC(=C(C=C1)N)Cl | |
141.6 | |
141.6 | |
Liquid |
97.5 | |
7°C | |
1.1510g/mL | |
99°C | |
97.5% min. (GC) | |
ClC6H3(CH3)NH2 | |
MFCD00007666 | |
1.151 | |
XGYLSRFSXKAYCR-UHFFFAOYSA-N | |
2-chloro-4-methylaniline | |
12007 | |
98% | |
2-Chloro-4-methylaniline |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin t
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 210-440-7
Recommended Storage : Product may darken during storage
- 2-Chloro-4-methylaniline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only