missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-4-methylaniline, 98%

Product Code. 10113400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10113400 25 mL Glass bottle 25mL
10427392 5 mL Glass bottle 5mL
10289503 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10113400 Supplier Thermo Scientific Acros Supplier No. 154670250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 615-65-6 | C7H8ClN | 141.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-65-6
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.6
MDL Number MFCD00007666
InChI Key XGYLSRFSXKAYCR-UHFFFAOYSA-N
Synonym 2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene
PubChem CID 12007
IUPAC Name 2-chloro-4-methylaniline
SMILES CC1=CC(=C(C=C1)N)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 7°C
Color Brown to Yellow
Density 1.1510g/mL
Boiling Point 223°C to 225°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(CH3)NH2
Refractive Index 1.5740 to 1.576
Quantity 25 mL
Beilstein 12, 989
Specific Gravity 1.151
Formula Weight 141.6
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-methylaniline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-4-methylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.