missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylaniline, 98%

Product Code. p-7026745
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11449997 5 g 5g
11459997 25 g 25g
11469997 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11449997

Brand: Thermo Scientific Alfa Aesar B22935.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 615-65-6 | C7H8ClN | 141.598 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 615-65-6
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00007666
InChI Key XGYLSRFSXKAYCR-UHFFFAOYSA-N
Synonym 2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene
PubChem CID 12007
IUPAC Name 2-chloro-4-methylaniline
SMILES CC1=CC(=C(C=C1)N)Cl

Specifikationer

Melting Point 7°C
Density 1.151
Boiling Point 223°C to 225°C
Flash Point 99°C (210°F)
Refractive Index 1.575
Quantity 5 g
UN Number UN3429
Beilstein 774514
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-methylaniline
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.