missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-4-methyl-5-nitropyridine, 97%

Product Code. 15466508
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466508 250 mg 250mg
15456508 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466508 Supplier Alfa Aesar™ Supplier No. H60039.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 23056-33-9
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
MDL Number MFCD00010688
InChI Key HWZUMEVIIGNXGM-UHFFFAOYSA-N
Synonym 2-chloro-5-nitro-4-picoline, 2-chloro-4-methyl-5-nitro-pyridine, 2-chloro-4-methyl-5-nitro pyridine, 2-chloro-5-nitro-4-methylpyridine, pyridine, 2-chloro-4-methyl-5-nitro, pubchem6223, acmc-209g0j, ksc201s3j, tpc-py010
PubChem CID 345364
IUPAC Name 2-chloro-4-methyl-5-nitropyridine
SMILES CC1=CC(Cl)=NC=C1[N+]([O-])=O

Specifications

Melting Point 37°C to 39°C
Boiling Point 91°C
Flash Point 113°C (235°F)
Quantity 250 mg
Formula Weight 172.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-methyl-5-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.