missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxybenzeneboronic acid, 95%, Thermo Scientific Chemicals

Product Code. 11491480
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491480 5 g 5g
11481480 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491480 Supplier Thermo Scientific Alfa Aesar Supplier No. H26917.06
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 219735-99-6 | Molecular Formula: C7H8BClO3 | Molecular Weight: 186.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 219735-99-6
Molecular Formula C7H8BClO3
Molecular Weight (g/mol) 186.40
MDL Number MFCD03411935
InChI Key LOCGPWGCRVKCFN-UHFFFAOYSA-N
Synonym 2-chloro-4-methoxyphenyl boronic acid, 2-chloro-4-methoxybenzeneboronic acid, 4-borono-3-chloroanisole, 2-chloro-4-methoxyphenyl-boronic acid, boronic acid, 2-chloro-4-methoxyphenyl, 2-chloro-4-methoxyphenylboronicacid, pubchem1781, 2-chloro-4-methoxy-phenyl boronic acid, acmc-209fq2, ksc201q2h
PubChem CID 2773330
IUPAC Name (2-chloro-4-methoxyphenyl)boronic acid
SMILES COC1=CC=C(B(O)O)C(Cl)=C1

Especificaciones

Melting Point ∼202°C
Quantity 5 g
Formula Weight 186.4
Percent Purity 95%
Chemical Name or Material 2-Chloro-4-methoxybenzeneboronic acid

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.