missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-hydroxybenzaldehyde, 97%

Product Code. 15462118
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462118 5 g 5g
15452118 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462118 Supplier Thermo Scientific Alfa Aesar Supplier No. H55771.06
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56962-11-9 | C7H5ClO2 | 156.565 g/mol

2-Chloro-4-hydroxybenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-hydroxybenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in organic solvents.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56962-11-9
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.565
MDL Number MFCD00052184
InChI Key ZMOMCILMBYEGLD-UHFFFAOYSA-N
Synonym benzaldehyde, 2-chloro-4-hydroxy, 2-chloro-4-hydroxy-benzaldehyde, benzaldehyde, chloro-4-hydroxy, 2-chloro-4-hydroxybenzaldehyde, pubchem7366, acmc-209lv6, ksc497o6r, 2-chlor-4-hydroxy-benzaldehyd, 2-chloranyl-4-oxidanyl-benzaldehyde
PubChem CID 185363
IUPAC Name 2-chloro-4-hydroxybenzaldehyde
SMILES C1=CC(=C(C=C1O)Cl)C=O

Specifications

Melting Point 145°C to 147°C
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in organic solvents.
Formula Weight 156.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-hydroxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.