missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-chloro-4-fluorobenzyl bromide, 99%, Thermo Scientific™
Click to view available options
Quantity:
2.5 g
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Molecular Formula | C7H5BrClF |
Linear Formula | F(Cl)C6H3CH2Br |
Quantity | 2.5 g |
Molecular Weight (g/mol) | 223.469 |
Formula Weight | 223.47 |
Show More |
Safety and Handling
Product Identifier
- 2-Chloro-4-fluorobenzyl bromide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'