missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzyl bromide, 97%

Product Code. 11495587
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495587 5 g 5g
11405597 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495587 Supplier Thermo Scientific Alfa Aesar Supplier No. B23565.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 45767-66-6 | C7H5BrClF | 223.469 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 45767-66-6
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.469
MDL Number MFCD00236025
InChI Key GAUUDQVOPUKGJD-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzyl bromide, 1-bromomethyl-2-chloro-4-fluorobenzene, 2-chloro-4-fluorobenzylbromide, alpha-bromo-2-chloro-4-fluorotoluene, benzene, 1-bromomethyl-2-chloro-4-fluoro, 2-choro-4-fluorobenzylbromide, pubchem4893, 1-bromomethyl-2-chloro-4-fluoro-benzene, ksc493m8r, 2-chloro-4-flurobenzyl bromide
PubChem CID 2725062
IUPAC Name 1-(bromomethyl)-2-chloro-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)Cl)CBr

Specifications

Melting Point 35°C to 39°C
Quantity 5 g
UN Number UN3261
Beilstein 3539265
Formula Weight 223.48
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorobenzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.