missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzoic acid, 99%, Thermo Scientific™

Product Code. 10172951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10172951 5 g Glass bottle 5g
10031861 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10172951 Supplier Thermo Scientific Acros Supplier No. 347340050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Chemical Identifiers

CAS 2252-51-9
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.56
MDL Number MFCD00010615
InChI Key GRPWQLDSGNZEQE-UHFFFAOYSA-N
Synonym benzoic acid, 2-chloro-4-fluoro, 2-chloro-4-fluoro-benzoic acid, 2-chloro-4-fluor obenzoic acid, pubchem1370, acmc-209fx5, ksc207e4n, rarechem al bo 0405, 2-chloro 4-fluoro benzoic acid, 2-chloro-4-fluoro benzoic acid, 4-fluoro-2-chloro-benzoic acid
PubChem CID 75259
IUPAC Name 2-chloro-4-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)Cl)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 183°C to 186°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(F)CO2H
Quantity 5 g
Formula Weight 174.56
Percent Purity 99%
Physical Form Powder or Flakes
Chemical Name or Material 2-Chloro-4-fluorobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-4-fluorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.