missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoroanisole, 97%, Thermo Scientific™

Catalog Number 11426197
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2267-25-6
C7H6ClFO
160.57
MFCD00672965
RKCGJVGMRPKPNY-UHFFFAOYSA-N
2-chloro-4-fluoroanisole, benzene, 2-chloro-4-fluoro-1-methoxy, 2-chloro-4-fluoro-1-methoxy-benzene, pubchem2658, acmc-1cnak, intermediates-zcf02609, ksc497i7l
2773580
2-chloro-4-fluoro-1-methoxybenzene
COC1=CC=C(F)C=C1Cl
This item is not returnable. View return policy
2267-25-6
C7H6ClFO
160.57
MFCD00672965
RKCGJVGMRPKPNY-UHFFFAOYSA-N
2-chloro-4-fluoroanisole, benzene, 2-chloro-4-fluoro-1-methoxy, 2-chloro-4-fluoro-1-methoxy-benzene, pubchem2658, acmc-1cnak, intermediates-zcf02609, ksc497i7l
2773580
2-chloro-4-fluoro-1-methoxybenzene
COC1=CC=C(F)C=C1Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.29
Boiling Point 197°C to 200°C
Flash Point 76°C (170°F)
Refractive Index 1.519
Quantity 5 g
Formula Weight 160.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluoroanisole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'