missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoroaniline, 97%

Product Code. 11422877
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422877 100 g 100g
11402877 5 g 5g
11412877 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422877 Supplier Thermo Scientific Alfa Aesar Supplier No. A15492.22
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2106-02-7 | C6H5ClFN | 145.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2106-02-7
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD00042530

Specifications

Density 1.219
Quantity 100 g
Odor Characteristic
Refractive Index 1.554
UN Number UN2810
Beilstein 2802559
Synonym 2-fluoro-3-chloroaniline, benzenamine, 3-chloro-2-fluoro, 3-chloro-2-fluorophenylamine, pubchem1529, 3,4-chlorofluoroaniline, 3-chloro-2 fluoroaniline, 2-fluoro-3-chloro-aniline, 3-chloro-2-fluoro aniline, 3-chloro-2-fluoro-aniline, ksc493k4l
Boiling Point 191°C to 192°C
InChI Key XWBTZHDDWRNOQH-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)Cl)F)N
IUPAC Name 3-chloro-2-fluoroaniline
PubChem CID 75014
Formula Weight 145.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluoroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.