missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4'-fluoroacetophenone, 99%, Thermo Scientific™

Product Code. 10266060 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10266060 100 g Glass bottle 100g
10794651 25 g Glass bottle 25g
10548873 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10266060 Supplier Thermo Scientific Acros Supplier No. 158171000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 456-04-2
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.59
MDL Number MFCD00011652
InChI Key UJZWJOQRSMOFMA-UHFFFAOYSA-N
Synonym 2-chloro-4'-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethanone, p-fluorophenacyl chloride, 4-fluorophenacyl chloride, alpha-chloro-4-fluoroacetophenone, 4-chloroacetyl-1-fluorobenzene, alpha-chloro-p-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethan-1-one, ethanone, 2-chloro-1-4-fluorophenyl, 2'-chloro-4-fluoroacetophenone
PubChem CID 120248
IUPAC Name 2-chloro-1-(4-fluorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CCl)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 47°C to 50°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula FC6H4COCH2Cl
Quantity 100 g
Formula Weight 172.59
Percent Purity 99%
Physical Form Flakes or Platelets
Chemical Name or Material 2-Chloro-4'-fluoroacetophenone
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-4'-fluoroacetophenone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.