missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoro-5-nitrotoluene, 97%, Thermo Scientific Chemicals

Product Code. p-7054824
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15449838 1 g 1g
15459838 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449838 Supplier Thermo Scientific Alfa Aesar Supplier No. H63244.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2-Chloro-4-fluoro-5-nitrotoluene, is used as a pharmaceutical intermediate. It is also used as Ceritinib intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-fluoro-5-nitrotoluene, is used as a pharmaceutical intermediate. It is also used as Ceritinib intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112108-73-3
Molecular Formula C7H5ClFNO2
Molecular Weight (g/mol) 189.57
MDL Number MFCD11110549
InChI Key YXVJHZWHPLOEAP-UHFFFAOYSA-N
Synonym 2-chloro-4-fluoro-5-nitrotoluene, 1-chloro-5-fluoro-2-methyl-4-nitro-benzene, benzene, 1-chloro-5-fluoro-2-methyl-4-nitro, acmc-2099cn, ksc682i1p, 2-chloro-4fluoro-5-nitrotoluene, 1-chloranyl-5-fluoranyl-2-methyl-4-nitro-benzene
PubChem CID 10103979
IUPAC Name 1-chloro-5-fluoro-2-methyl-4-nitrobenzene
SMILES CC1=CC(=C(C=C1Cl)F)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 1 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 189.57
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Chloro-4-fluoro-5-nitrotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.