missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoro-1-iodobenzene, 98%

Product Code. p-7081843
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11489852

Brand: Thermo Scientific Alfa Aesar A12010.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 101335-11-9 | C6H3ClFI | 256.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101335-11-9
Molecular Formula C6H3ClFI
Molecular Weight (g/mol) 256.44
MDL Number MFCD00051774
InChI Key POTCKVPDYXEGSV-UHFFFAOYSA-N
Synonym 2-chloro-4-fluoroiodobenzene, 1-chloro-5-fluoro-2-iodobenzene, benzene, 2-chloro-4-fluoro-1-iodo, 2-chloro-4-fluoro-1-iodo-benzene, pubchem3434, acmc-1bu0g, ksc494m4h, 4-fluoro-2-chloro-iodobenzene, 2-chloro-4-fluoro-iodobenzene, 2-chloro-4-fluoro-1-iodobenzol
PubChem CID 2736559
IUPAC Name 2-chloro-4-fluoro-1-iodobenzene
SMILES FC1=CC=C(I)C(Cl)=C1

Specifications

Boiling Point 96°C to 98°C (15 mmHg)
Refractive Index 1.601
Quantity 5 g
Beilstein 2435080
Sensitivity Light sensitive
Formula Weight 256.45
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-fluoro-1-iodobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.