missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoro-1-iodobenzene, 97%, Thermo Scientific™

Product Code. p-4391250
Click to view available options
Quantity:
50 g
100 g
Packaging:
Amber Glass Bottle
Unit Size:
100g
50g
This item is not returnable. View return policy

Product Code. 10271003

Brand: Thermo Scientific Maybridge TL00223EE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 101335-11-9
Molecular Formula C6H3ClFI
Molecular Weight (g/mol) 256.44
MDL Number MFCD00051774
InChI Key POTCKVPDYXEGSV-UHFFFAOYSA-N
Synonym 2-chloro-4-fluoroiodobenzene, 1-chloro-5-fluoro-2-iodobenzene, benzene, 2-chloro-4-fluoro-1-iodo, 2-chloro-4-fluoro-1-iodo-benzene, pubchem3434, acmc-1bu0g, ksc494m4h, 4-fluoro-2-chloro-iodobenzene, 2-chloro-4-fluoro-iodobenzene, 2-chloro-4-fluoro-1-iodobenzol
PubChem CID 2736559
IUPAC Name 2-chloro-4-fluoro-1-iodobenzene
SMILES FC1=CC=C(I)C(Cl)=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 50 g
Formula Weight 256.44
Percent Purity 97%
Chemical Name or Material 2-chloro-4-fluoro-1-iodobenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.