missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(dimethylamino)benzaldehyde, ≥95%, Thermo Scientific™

Product Code. p-2698041
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10530553 1 g Amber Glass Bottle 1g
10243092 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10530553 Supplier Thermo Scientific Maybridge Supplier No. SEW05671DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1424-66-4
Molecular Formula C9H10ClNO
Molecular Weight (g/mol) 183.64
MDL Number MFCD00021051
InChI Key XSQFAWMDRFSIMY-UHFFFAOYSA-N
Synonym 2-chloro-4-dimethylamino benzaldehyde, benzaldehyde, 2-chloro-4-dimethylamino, benzaldehyde,2-chloro-4-dimethylamino, 2-chloro-4-n,n-bis methyl amino benzaldehyde, 4-dimethylamino-2-chlorobenzaldehyde, acmc-209coh, 3-chloro-n,n-dimethyl-4-formylaniline
PubChem CID 74021
IUPAC Name 2-chloro-4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C(Cl)=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3N(CH3)C6H3(Cl)CHO
Quantity 1 g
Formula Weight 183.64
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(dimethylamino)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.