missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-cyanopyridine, 97%, Thermo Scientific Chemicals

Catalog Number 11379855
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
33252-30-1
C6H3ClN2
138.554
MFCD00174318
QRXBTPFMCTXCRD-UHFFFAOYSA-N
2-chloro-4-cyanopyridine, 2-chloroisonicotinonitrile, 2-chloro-4-pyridinecarbonitrile, 2-chloro-isonicotinonitrile, 4-cyano-2-chloropyridine, 4-pyridinecarbonitrile, 2-chloro, 2-chloro-4-cyano pyridine, 2-chlorisonicotinonitril, pubchem2582
568638
2-chloropyridine-4-carbonitrile
C1=CN=C(C=C1C#N)Cl
This item is not returnable. View return policy
33252-30-1
C6H3ClN2
138.554
MFCD00174318
QRXBTPFMCTXCRD-UHFFFAOYSA-N
2-chloro-4-cyanopyridine, 2-chloroisonicotinonitrile, 2-chloro-4-pyridinecarbonitrile, 2-chloro-isonicotinonitrile, 4-cyano-2-chloropyridine, 4-pyridinecarbonitrile, 2-chloro, 2-chloro-4-cyano pyridine, 2-chlorisonicotinonitril, pubchem2582
568638
2-chloropyridine-4-carbonitrile
C1=CN=C(C=C1C#N)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 69°C to 72°C
Boiling Point 104°C to 106°C (15 mmHg)
Quantity 5 g
UN Number UN3439
Beilstein 113927
Formula Weight 138.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-cyanopyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'