missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-aminopyridine, 97%

Product Code. 10246873 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10246873 100 g Glass bottle 100g
10594911 5 g Glass bottle 5g
10041871 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10246873 Supplier Thermo Scientific Acros Supplier No. 345451000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14432-12-3 | C5H5ClN2 | 128.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14432-12-3
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
InChI Key BLBDTBCGPHPIJK-UHFFFAOYSA-N
Synonym 4-amino-2-chloropyridine, 2-chloro-4-aminopyridine, 2-chloro-pyridin-4-ylamine, 4-pyridinamine, 2-chloro, 2-chloro-4-pyridinamine, 2-chlorpyridin-4-amin, 4-amino-6-chloropyridine, 2-chloro-4-pyridylamine, 2-chloro-4-amino pyridine, 4-amino-2-chloro pyridine
PubChem CID 84432
IUPAC Name 2-chloropyridin-4-amine
SMILES C1=CN=C(C=C1N)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 92°C to 94°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 g
Formula Weight 128.56
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-aminopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-4-aminopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.