missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-dimethylpyrimidine, 97%

Product Code. 11976551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11976551 25 g 25g
11966551 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11976551 Supplier Thermo Scientific Acros Supplier No. 442550250
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4472-44-0 | C6H7ClN2 | 142.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4472-44-0
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
InChI Key RZVPFDOTMFYQHR-UHFFFAOYSA-N
Synonym pyrimidine, 2-chloro-4,6-dimethyl, 2-chloro-4,6-dimethyl-pyrimidine, pubchem6904, acmc-1adw7, 5-23-05-00401 beilstein handbook reference, ksc495i5r, 2-chlor-4,6-dimethylpyrimidine, 4,6-dimethyl-2-chloropyrimidine, 2-chloro-4,6-dimethyl-1,3-diazine, 2-chloro-4,6-dimethylpyrimidine
PubChem CID 20550
IUPAC Name 2-chloro-4,6-dimethylpyrimidine
SMILES CC1=CC(=NC(=N1)Cl)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 36°C to 38°C
Color White to Yellow
Boiling Point 210°C to 212°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 25 g
Formula Weight 142.59
Percent Purity 97%
Physical Form Powder, Crystalline Powder or Low Melting Solid
Chemical Name or Material 2-Chloro-4, 6-dimethylpyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-4,6-dimethylpyrimidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.