missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%

Product Code. 10114630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10114630 25 g Glass bottle 25g
10454463 5 g Glass bottle 5g
11906581 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10114630 Supplier Thermo Scientific Acros Supplier No. 263000250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3140-73-6 | C5H6ClN3O2 | 175.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3140-73-6
Molecular Formula C5H6ClN3O2
Molecular Weight (g/mol) 175.57
MDL Number MFCD00075607
InChI Key GPIQOFWTZXXOOV-UHFFFAOYSA-N
Synonym cdmt, 1,3,5-triazine, 2-chloro-4,6-dimethoxy, 2-chloro-4,6-dimethoxy-s-triazine, 2-chloro-4,6-dimethoxy-1,3,5 triazine, pubchem20734, acmc-1cnh7, ksc567a8j, 2-chloro-4,6-dimethoxy s-triazine, 1,5-triazine, 2-chloro-4,6-dimethoxy
PubChem CID 18450
IUPAC Name 2-chloro-4,6-dimethoxy-1,3,5-triazine
SMILES COC1=NC(OC)=NC(Cl)=N1

Specifications

Melting Point 74.0°C to 78.0°C
Color White
Boiling Point 99.0°C to 100.0°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents,sparingly soluble in light petroleum and ethanol
Formula Weight 175.57
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4,6-dimethoxy-1,3,5-triazine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-4,6-dimethoxy-1,3,5-triazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.