missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,5-difluorobenzoic acid, 99+%, Thermo Scientific™

Product Code. 10482061 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10482061 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10482061 Supplier Thermo Scientific Acros Supplier No. 277690050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 110877-64-0
CAS Min % 99.0
CAS Max % 100.0
Melting Point 103.0°C to 106.0°C
Color White
Boiling Point 258.0°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C7H2ClF2O2
Linear Formula ClC6H2(COOH)F2
MDL Number MFCD00077479
Quantity 5 g
Synonym 2-chloro-4,5-difluoro-benzoic acid, 2'-chloro-4',5'-difluorobenzoic acid, benzoic acid, 2-chloro-4,5-difluoro, pubchem1339, ksc177o4l, rarechem al bo 1000, timtec-bb sbb003616, 2-chloro-4,5-difluorobenzoic acid, benzoicacid, 2-chloro-4,5-difluoro
Solubility Information Solubility in water: 5.0g/L (20 c)
InChI Key CGFMLBSNHNWJAW-UHFFFAOYSA-M
SMILES [O-]C(=O)C1=CC(F)=C(F)C=C1Cl
IUPAC Name 2-chloro-4,5-difluorobenzoic acid
Molecular Weight (g/mol) 191.54
PubChem CID 737171
Formula Weight 192.55
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Chloro-4, 5-difluorobenzoic acid, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-4,5-difluorobenzoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.