missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-[2-(4-fluorophenyl)ethyl]-6-methylpyrimidine, 97%, Thermo Scientific™

Catalog Number 15418176
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
372183-70-5
C13H12ClFN2
250.70
MFCD04974533
VALSYGYSZKIHMQ-UHFFFAOYSA-N
2-chloro-4-2-4-fluorophenyl ethyl-6-methylpyrimidine, 2-chloro-4-2-4-fluorophenyl-ethyl-6-methylpyrimidine, c13h13clfn3, 2-chloro-4-4-fluorophenethyl-6-methylpyrimidine
53411892
2-chloro-4-[2-(4-fluorophenyl)ethyl]-6-methylpyrimidine
CC1=CC(CCC2=CC=C(F)C=C2)=NC(Cl)=N1
This item is not returnable. View return policy
372183-70-5
C13H12ClFN2
250.70
MFCD04974533
VALSYGYSZKIHMQ-UHFFFAOYSA-N
2-chloro-4-2-4-fluorophenyl ethyl-6-methylpyrimidine, 2-chloro-4-2-4-fluorophenyl-ethyl-6-methylpyrimidine, c13h13clfn3, 2-chloro-4-4-fluorophenethyl-6-methylpyrimidine
53411892
2-chloro-4-[2-(4-fluorophenyl)ethyl]-6-methylpyrimidine
CC1=CC(CCC2=CC=C(F)C=C2)=NC(Cl)=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 250.07
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Chloro-4-[2-(4-fluorophenyl)ethyl]-6-methylpyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'