missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-(trifluoromethyl)pyridine, 98%, Thermo Scientific™

Product Code. 11308095
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11308095 1 g 1g
11318095 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308095 Supplier Thermo Scientific Alfa Aesar Supplier No. L08303.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65753-47-1
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.542
MDL Number MFCD00042223
InChI Key RXATZPCCMYMPME-UHFFFAOYSA-N
Synonym 2-chloro-3-trifluoromethyl pyridine, 2-chloro-3-trifluoro methylpyridine, pyridine, 2-chloro-3-trifluoromethyl, 2,3-ctf, 2-chloro-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-chloropyridine, zlchem 248, pubchem2399, acmc-1bejp, ksc352q1t
PubChem CID 589833
IUPAC Name 2-chloro-3-(trifluoromethyl)pyridine
SMILES C1=CC(=C(N=C1)Cl)C(F)(F)F

Specifications

Melting Point 37°C to 40°C
Boiling Point 166°C to 168°C
Quantity 1 g
UN Number UN2923
Beilstein 4179716
Formula Weight 181.55
Percent Purity 98%
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.