missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. 11350716 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11350716 25 g Glass bottle 25g
10567600 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11350716 Supplier Thermo Scientific Acros Supplier No. 241790250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 65753-47-1
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.54
InChI Key RXATZPCCMYMPME-UHFFFAOYSA-N
Synonym 2-chloro-3-trifluoromethyl pyridine, 2-chloro-3-trifluoro methylpyridine, pyridine, 2-chloro-3-trifluoromethyl, 2,3-ctf, 2-chloro-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-chloropyridine, zlchem 248, pubchem2399, acmc-1bejp, ksc352q1t
PubChem CID 589833
IUPAC Name 2-chloro-3-(trifluoromethyl)pyridine
SMILES C1=CC(=C(N=C1)Cl)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 36.0°C to 40.0°C
Boiling Point 166.0°C to 168.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: reacts at high temperature with strong acids and,alkalis
Formula Weight 181.54
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material 2-chloro-3-(trifluoromethyl)pyridine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-3-(trifluoromethyl)pyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.