missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-3-picoline, 98%, Thermo Scientific™

Product Code. 10289573 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10289573 100 g Glass bottle 100g
10411913 5 g Glass bottle 5g
10270303 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10289573 Supplier Thermo Scientific Acros Supplier No. 379581000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 18368-76-8
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00234177
InChI Key RKVUCIFREKHYTL-UHFFFAOYSA-N
PubChem CID 87603
IUPAC Name 2-chloro-3-methylpyridine
SMILES CC1=C(N=CC=C1)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 192°C to 193°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5320 to 1.534
Quantity 100 g
Specific Gravity 1.17
Synonym 2-chloro-3-picoline, 2-chloro 3-methylpyridine, 3-picoline, 2-chloro, pyridine, 2-chloro-3-methyl, 2-chloro-3-methyl-pyridine, pyridine,2-chloro-3-methyl, pubchem2074, 2-chloro-methyl-pyridine, 2-chloro3-methyl-pyridine
Formula Weight 127.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloro-3-picoline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-3-picoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.