missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-chloro-3-picoline, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 18368-76-8 |
---|---|
Molecular Formula | C6H6ClN |
Molecular Weight (g/mol) | 127.57 |
MDL Number | MFCD00234177 |
InChI Key | RKVUCIFREKHYTL-UHFFFAOYSA-N |
PubChem CID | 87603 |
IUPAC Name | 2-chloro-3-methylpyridine |
SMILES | CC1=C(N=CC=C1)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10411913
|
Thermo Scientific Acros
379580050 |
5 g | Glass bottle |
N/A
|
N/A | ||||
10270303
|
Thermo Scientific Acros
379580250 |
25 g | Glass bottle |
N/A
|
N/A | ||||
10289573
|
Thermo Scientific Acros
379581000 |
100 g | Glass bottle |
N/A
|
N/A | ||||
Description
Chemical Identifiers
18368-76-8 | |
127.57 | |
RKVUCIFREKHYTL-UHFFFAOYSA-N | |
2-chloro-3-methylpyridine |
C6H6ClN | |
MFCD00234177 | |
87603 | |
CC1=C(N=CC=C1)Cl |
Specifications
18368-76-8 | |
100.0 | |
1.1700g/mL | |
79°C | |
97.5% min. (GC) | |
1.5320 to 1.534 | |
1.17 | |
RKVUCIFREKHYTL-UHFFFAOYSA-N | |
2-chloro-3-methylpyridine | |
87603 | |
98% | |
2-Chloro-3-picoline |
97.5 | |
Colorless to Yellow | |
192°C to 193°C | |
Authentic | |
C6H6ClN | |
MFCD00234177 | |
2-chloro-3-picoline, 2-chloro 3-methylpyridine, 3-picoline, 2-chloro, pyridine, 2-chloro-3-methyl, 2-chloro-3-methyl-pyridine, pyridine,2-chloro-3-methyl, pubchem2074, 2-chloro-methyl-pyridine, 2-chloro3-methyl-pyridine | |
CC1=C(N=CC=C1)Cl | |
127.57 | |
127.57 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 242-242-1
Recommended Storage : this product may darken during storage
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Chloro-3-picoline
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only