missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-(pentafluoroethyl)pyridine, 96%, Thermo Scientific™

Product Code. 15462719
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1816284-10-2
Color Yellow
Molecular Formula C7H3ClF5N
Quantity 100 mg
InChI Key SXJAZKPSUJJAKV-UHFFFAOYSA-N
SMILES C1=CC(=C(N=C1)Cl)C(C(F)(F)F)(F)F
IUPAC Name 2-chloro-3-(1,1,2,2,2-pentafluoroethyl)pyridine
Molecular Weight (g/mol) 231.55
PubChem CID 99728609
Formula Weight 231.55
Percent Purity 96%
Physical Form Viscous Liquid
Chemical Name or Material 2-Chloro-3-(pentafluoroethyl)pyridine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'