missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-nitropyridine, 99+%

Product Code. 10197563 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10197563 100 g Glass bottle 100g
10705371 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10197563 Supplier Thermo Scientific Acros Supplier No. 109731000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5470-18-8 | C5H3ClN2O2 | 158.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5470-18-8
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.54
MDL Number MFCD00006232
InChI Key UUOLETYDNTVQDY-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-3-nitro, 3-nitro-2-chloropyridine, 2-chloro-3-nitro-pyridine, 2-chloro-3-nitro pyridine, 2-chlor-3-nitropyridin, zlchem 303, pubchem1229, 2-chloro-nitropyridine, 2-chloro-3nitropyridine, 2-chloro3-nitropyridine
PubChem CID 79613
IUPAC Name 2-chloro-3-nitropyridine
SMILES C1=CC(=C(N=C1)Cl)[N+](=O)[O-]

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 100.0°C to 103.0°C
Color Yellow
Density 185g/mL
Flash Point 185°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Linear Formula Cl(C5H3N)NO2
Refractive Index 1.4540 to 1.4560
Quantity 100 g
Beilstein 20, I, 82
Solubility Information Solubility in water: insoluble
Formula Weight 158.54
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloro-3-nitropyridine
Show More Show Less
compliance-icons
Identifiant produit
  • 2-Chloro-3-nitropyridine
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.