missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-cyano-6-methylpyridine, 99%

Product Code. 11477557
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11477557 5 g 5g
11467557 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11477557

Brand: Thermo Scientific Alfa Aesar A15648.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 28900-10-9 | C7H5ClN2 | 152.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28900-10-9
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00082767
InChI Key YSBNBAYNISAUIT-UHFFFAOYSA-N
Synonym 2-chloro-6-methylnicotinonitrile, 2-chloro-3-cyano-6-methylpyridine, 2-chloro-6-methyl-3-pyridinecarbonitrile, 2-chloro-3-cyano-6-picoline, 3-pyridinecarbonitrile, 2-chloro-6-methyl, 2-chloro-6-methyl-nicotinonitrile, pubchem6128, 2-chloro-6-methylnicotinonitrile #, 2-chloro-3-cyano-6-methyl pyridine, 2-chloro-3-cyano-6-methyl-pyridine
PubChem CID 520400
IUPAC Name 2-chloro-6-methylpyridine-3-carbonitrile
SMILES CC1=CC=C(C#N)C(Cl)=N1

Specifications

Melting Point 114°C to 117°C
Quantity 5 g
Beilstein 120414
Formula Weight 152.58
Percent Purity 99%
Chemical Name or Material 2-Chloro-3-cyano-6-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.