missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,6-difluorophenylacetic acid, 97%, Thermo Scientific™

Catalog Number 15473537
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
261762-52-1
C8H5ClF2O2
206.57
MFCD01631448
LITZKYPXFPYSOE-UHFFFAOYSA-N
2-chloro-3,6-difluorophenylacetic acid, 2-2-chloro-3,6-difluorophenyl acetic acid, 2-chloro-3,6-difluorophenyl acetic acid, 2-chloro-3,6-difluoro-phenyl-acetic acid, pubchem2707, 2-chloro-3,6-difluorophenyl-acetic acid
2773566
2-(2-chloro-3,6-difluorophenyl)acetic acid
OC(=O)CC1=C(F)C=C(Cl)C=C1F
This item is not returnable. View return policy
261762-52-1
C8H5ClF2O2
206.57
MFCD01631448
LITZKYPXFPYSOE-UHFFFAOYSA-N
2-chloro-3,6-difluorophenylacetic acid, 2-2-chloro-3,6-difluorophenyl acetic acid, 2-chloro-3,6-difluorophenyl acetic acid, 2-chloro-3,6-difluoro-phenyl-acetic acid, pubchem2707, 2-chloro-3,6-difluorophenyl-acetic acid
2773566
2-(2-chloro-3,6-difluorophenyl)acetic acid
OC(=O)CC1=C(F)C=C(Cl)C=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 135°C to 139°C
Quantity 250 mg
Formula Weight 206.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-3,6-difluorophenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'