missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,5-dinitrobenzoic acid, 97% (dry wt.), may cont. up to ca 5% water

Product Code. 11399649
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399649 25 g 25g
11437617 100 g 100g
11447617 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399649 Supplier Thermo Scientific Alfa Aesar Supplier No. A15675.14
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2497-91-8 | C7H3ClN2O6 | 246.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2497-91-8
Molecular Formula C7H3ClN2O6
Molecular Weight (g/mol) 246.56
MDL Number MFCD00007070
InChI Key ADTKEYLCJYYHHH-UHFFFAOYSA-N
Synonym 2-chloro-3,5-dinitrobenzoic acid, benzoic acid, 2-chloro-3,5-dinitro, 2-carboxy-4,6-dinitrochlorobenzene, 3,5-dinitro-2-chlorobenzoic acid, acmc-209uud, timtec-bb sbb003177, wln: wnr bg cvq enw, 4-09-00-01359 beilstein handbook reference, ksc495o8b, 2-chloro-3,5-dinitro-benzoicaci
PubChem CID 17247
IUPAC Name 2-chloro-3,5-dinitrobenzoic acid
SMILES OC(=O)C1=CC(=CC(=C1Cl)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 196°C to 201°C
Quantity 25 g
Beilstein 2146480
Formula Weight 246.56
Percent Purity 97%
Assay (dry wt.), may cont. up to ≈5% water
Chemical Name or Material 2-Chloro-3,5-dinitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.