missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,5-dibromo-6-methylpyridine, Thermo Scientific™

Product Code. 10585080
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10585080 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10585080 Supplier Thermo Scientific Maybridge Supplier No. MAY00204DA
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1000018-58-5
Packaging Amber Glass Bottle
Molecular Formula C6H4Br2ClN
MDL Number MFCD09702430
Quantity 1 g
Synonym 2-chloro-3,5-dibromo-6-methylpyridine, acmc-2097lo
InChI Key AYZAQZJMQIXSLF-UHFFFAOYSA-N
SMILES CC1=NC(Cl)=C(Br)C=C1Br
IUPAC Name 3,5-dibromo-2-chloro-6-methylpyridine
Molecular Weight (g/mol) 285.36
PubChem CID 26966711
Formula Weight 285.37
Percent Purity 97%
Chemical Name or Material 2-chloro-3, 5-dibromo-6-methylpyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.