missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,4-dimethoxypyridine, 97%, Thermo Scientific Chemicals

Product Code. 15406557
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15406557 1 g 1g
15416557 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15406557 Supplier Thermo Scientific Alfa Aesar Supplier No. H33697.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Chloro-3,4-dimethoxypyridine is used in the preparation of 3, 4-dimethoxy-pyridine by using nickel(II) chloride hexahydrate and triphenylphosphine as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-3,4-dimethoxypyridine is used in the preparation of 3, 4-dimethoxy-pyridine by using nickel(II) chloride hexahydrate and triphenylphosphine as a catalyst.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101664-59-9
Molecular Formula C7H9Cl2NO2
Molecular Weight (g/mol) 210.054
MDL Number MFCD03265283
InChI Key AOBQXXVCWDAJFE-UHFFFAOYSA-N
Synonym 2-chloro-3,4-dimethoxypyridine hydrochloride, 2-chloro-3,4-dimethoxypyridine hcl, 2-chloro-3,4-dimethoxypyridine-hydrogen chloride 1/1
PubChem CID 24728279
IUPAC Name 2-chloro-3,4-dimethoxypyridine;hydrochloride
SMILES COC1=C(C(=NC=C1)Cl)OC.Cl

Specifications

Melting Point 37°C to 42°C
Quantity 1 g
Formula Weight 173.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-3,4-dimethoxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.