missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3',4'-dihydroxyacetophenone, 97%

Product Code. 11475497
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475497 25 g 25g
11485497 100 g 100g
11495497 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475497 Supplier Thermo Scientific Alfa Aesar Supplier No. B23496.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 99-40-1 | C8H7ClO3 | 186.591 g/mol

2-Chloro-3',4'-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-3′,4′-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.

Solubility
Soluble in DMSO, methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-40-1
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00002200
InChI Key LWTJEJCZJFZKEL-UHFFFAOYSA-N
Synonym 2-chloro-3',4'-dihydroxyacetophenone, 2-chloro-1-3,4-dihydroxyphenyl ethanone, 4-chloroacetyl catechol, 3,4-dihydroxyphenacyl chloride, ethanone, 2-chloro-1-3,4-dihydroxyphenyl, unii-fn6949l56q, 3',4'-dihydroxy-2-chloroacetophenone, chloromethyl 3,4-dihydroxyphenyl ketone, 2-chloro-3,4-dihydroxyacetophenone, .alpha.-chloro-3,4-dihydroxyacetophenone
PubChem CID 66834
ChEBI CHEBI:51844
IUPAC Name 2-chloro-1-(3,4-dihydroxyphenyl)ethanone
SMILES C1=CC(=C(C=C1C(=O)CCl)O)O

Specifications

Melting Point 172°C to 176°C
Quantity 25 g
Beilstein 2092660
Solubility Information Soluble in DMSO,methanol.
Formula Weight 186.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-3',4'-dihydroxyacetophenone
compliance-icons
Product Identifier
  • 2-Chloro-3',4'-dihydroxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2-Chloro-3',4'-dihydroxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.