missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-(phenylhydrazono)acetic Acid Ethyl Ester, TRC

Product Code. 30364669
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364669 500 mg -
30358937 5 g -
30355753 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364669 Supplier Toronto Research Chemicals Supplier No. TRCC385953500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28663-68-5
Molecular Formula C10H11N2O2Cl
Molecular Weight (g/mol) 226.65
Synonym Ethyl (Phenylhydrazono)chloromethylcarboxylate, Ethyl 2-(2-Phenylhydrazono)-2-chloroacetate, Ethyl 2-Chloro-2-(phenylhydrazono)acetate, Ethyl Chloro(phenylhydrazono)acetate, Ethyl Chloroglyoxylate phenylhydrazone, NSC 333417, Oxalyl Chloride Phenylhydrazone Ethyl Ester
IUPAC Name ethyl (2E)-2-chloro-2-(phenylhydrazinylidene)acetate
SMILES CCOC(=O)\C(=N\Nc1ccccc1)\Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H11ClN2O2/c1-2-15-10(14)9(11)13-12-8-6-4-3-5-7-8/h3-7,12H,2H2,1H3/b13-9+
Formula Weight 226.0509
Chemical Name or Material 2-Chloro-2-(phenylhydrazono)acetic Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.