missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-methylpropane, TRC

Product Code. 30364865
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
Product Code. Quantity unitSize
30364865 25 mL 25mL
1 options
This item is not returnable. View return policy

Product Code. 30364865

Brand: Toronto Research Chemicals TRCC36968025ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloro-2-methylpropane
CAS 507-20-0
Molecular Formula C4 H9 Cl
Quantity 25 mL
Synonym Propane, 2-chloro-2-methyl- (8CI, 9CI, ACI), 2-Chloro-2-methylpropane (ACI), 1,1-Dimethylethyl chloride, 1-Chloro-1,1-dimethylethane, 2-Chloroisobutane, 2-Methyl-2-chloropropane, 2-Methyl-2-propyl chloride, Chlorotrimethylmethane, NSC 6527, tert-Butyl chloride, Trimethylchloromethane
InChI Formula InChI=1S/C4H9Cl/c1-4(2,3)5/h1-3H3
SMILES CC(C)(C)Cl
IUPAC Name 2-chloro-2-methylpropane
Molecular Weight (g/mol) 92.567
Formula Weight 92.039
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.