missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-methylbutane, TRC

Product Code. 30358737
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30358737 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30358737 Supplier Toronto Research Chemicals Supplier No. TRCC45562010ML
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material 2-Chloro-2-methylbutane
CAS 594-36-5
Molecular Formula C5H11Cl
Quantity 10 mL
Synonym 1,1-Dimethyl-chloropropane, 1,1-Dimethylpropyl Chloride, 2-Methyl-2-chlorobutane, NSC 7900, tert-Amyl Chloride, tert-Pentyl Chloride
InChI Formula InChI=1S/C5H11Cl/c1-4-5(2,3)6/h4H2,1-3H3
SMILES CCC(C)(C)Cl
IUPAC Name 2-chloro-2-methylbutane
Molecular Weight (g/mol) 106.59
Formula Weight 106.0549
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.