Learn More
2-Chloro-2',6'-dimethylacetanilide, 99%
Description
2-Chloro-2',6'-dimethylacetanilide is used in the determination of the enantiomers of tocainide in blood plasma by gas-liquid chromatography with electron-capture detection. It is a nonsteroidal antinflammatory drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in Chloroform, Methanol. Insoluble in water.
Notes
Keep the container tightly closed in a cool, dry and well ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 143°C to 148°C |
| Quantity | 50 g |
| Beilstein | 781309 |
| Solubility Information | Soluble in Chloroform,Methanol. Insoluble in water. |
| Formula Weight | 197.67 |
| Percent Purity | 99% |
| Chemical Name or Material | 2-Chloro-2',6'-dimethylacetanilide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.