missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2,2-difluoroacetophenone, 96%

Product Code. 11383856
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
missing translation for 'unitSize'
1g
5g
This item is not returnable. View return policy

Product Code. 11383856

missing translation for 'mfr': Thermo Scientific Alfa Aesar L16963.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 384-67-8 | C8H5ClF2O | 190.574 g/mol

Reagent is an effective product for the synthesis of difluoromethylated phenols in the presence of mild base and aqueous solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent is an effective product for the synthesis of difluoromethylated phenols in the presence of mild base and aqueous solvent.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 384-67-8
Molecular Formula C8H5ClF2O
Molecular Weight (g/mol) 190.574
MDL Number MFCD00042123
InChI Key MNOONJNILVDLSW-UHFFFAOYSA-N
Synonym 2-chloro-2,2-difluoroacetophenone, chlorodifluoroacetophenone, 2,2,2-chlorodifluoroacetophenone, 2-chloro-2,2-difluoro-1-phenylethan-1-one, ethanone, 2-chloro-2,2-difluoro-1-phenyl, zlchem 965, chlorodifluroacetophenone, acmc-1bn70, ksc226m9l, chlorodifluoromethyl phenyl ketone
PubChem CID 238296
IUPAC Name 2-chloro-2,2-difluoro-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(F)(F)Cl

Specifications

Density 1.293
Boiling Point 95°C to 97°C (37 mmHg)
Flash Point 72°C (161°F)
Refractive Index 1.497
Quantity 1 g
Beilstein 2251448
Solubility Information Difficult to mix.
Formula Weight 190.58
Percent Purity 96%
Chemical Name or Material 2-Chloro-2,2-difluoroacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.