missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-11-(1-piperazinyl)dibenzo[b,f]-1,4-oxazepine, 98%

Product Code. p-7042869
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15480337 250 mg 250mg
15460337 1 g 1g
15470337 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15480337 Supplier Thermo Scientific Alfa Aesar Supplier No. J64066.MD
Add to basket
This item is not returnable. View return policy

A tricyclic norepinephrine uptake inhibitor | CAS: 14028-44-5 | C17H16ClN3O | 313.785 g/mol

A tricyclic norepinephrine uptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14028-44-5
Molecular Formula C17H16ClN3O
Molecular Weight (g/mol) 313.785
MDL Number MFCD00069210
InChI Key QWGDMFLQWFTERH-UHFFFAOYSA-N
Synonym amoxapine, asendin, demolox, moxadil, amoxan, amoxapinum, amoxapina, amoxepine, asendis, desmethylloxapin
PubChem CID 2170
ChEBI CHEBI:2675
IUPAC Name 8-chloro-6-piperazin-1-ylbenzo[b][1,4]benzoxazepine
SMILES C1CN(CCN1)C2=NC3=CC=CC=C3OC4=C2C=C(C=C4)Cl

Specifications

Color White
Quantity 250 mg
Merck Index 14,576
Solubility Information Soluble in methanol,DMSO or chloroform.
Formula Weight 313.78
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Chloro-11-(1-piperazinyl)dibenzo[b,f]-1,4-oxazepine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.