missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-1-morpholinoethan-1-one, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 163.6 |
Percent Purity | 95% |
Chemical Name or Material | 2-Chloro-1-morpholinoethan-1-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'