Learn More
2-Chloro-1-methylpyridinium iodide, 97%
Description
2-Chloro-1-methylpyridinium iodide is a reagent in a wide variety of dehydrative coupling reactions. It is also used to form carboxylate esters from acids and alcohols, carboxamides from acids and amines, lactones from -hydroxy acids and carbodiimides from N,N-disubstituted thioureas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Chloro-1-methylpyridinium iodide is a reagent in a wide variety of dehydrative coupling reactions. It is also used to form carboxylate esters from acids and alcohols, carboxamides from acids and amines, lactones from -hydroxy acids and carbodiimides from N,N-disubstituted thioureas.
Solubility
Soluble in Methanol (50 mg/ml). Insoluble in water.
Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | ∼200°C (decomposition) |
| Quantity | 500 g |
| Beilstein | 3572320 |
| Sensitivity | Light and moisture sensitive |
| Merck Index | 14,6301 |
| Solubility Information | Soluble in Methanol (50mg/ml). Insoluble in water. |
| Formula Weight | 255.49 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Chloro-1-methylpyridinium iodide |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.