missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-methylimidazole-5-boronic acid pinacol ester, 95%

Product Code. 15407875
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 15407875

Brand: Thermo Scientific Alfa Aesar H31692.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1315281-29-8 | C10H16BClN2O2 | 242.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Sparingly soluble in water (0.23 g/L)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1315281-29-8
Melting Point 131°C to 135°C
Molecular Formula C10H16BClN2O2
MDL Number MFCD11100959
Quantity 250 mg
Synonym 2-chloro-1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-imidazole, 2-chloro-1-methylimidazole-5-boronic acid pinacol ester, 2-chloro-1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl imidazole, 2-chloro-1-methyl-1h-imidazole-5-boronic acid pinacol ester, 2-chloro-1-methyl-1h-imidazole-5-boronic acid, pinacol ester, 2-chloro-1-methyl-1h-imidazol-5-yl boronic acid pinacol ester, 2-chloro-1-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-imidazole, 2-chloro-1-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl imidazole
Solubility Information Sparingly soluble in water (0.23g/L)
InChI Key MOQXZCYTVMKINQ-UHFFFAOYSA-N
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N2C)Cl
IUPAC Name 2-chloro-1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)imidazole
Molecular Weight (g/mol) 242.51
PubChem CID 45588157
Formula Weight 242.52
Percent Purity 95%
Chemical Name or Material 2-Chloro-1-methylimidazole-5-boronic acid pinacol ester
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.