missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-iodo-4-nitrobenzene, 98%

Product Code. 11413947
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11413947

Brand: Thermo Scientific Alfa Aesar A19340.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41252-96-4 | C6H3ClINO2 | 283.449 g/mol

3-chloro-4-trifluoromethylanilinewas prepared in two steps by reaction of trifluoromethylcopper with 3-chloro-4-iodonitrobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-chloro-4-trifluoromethylanilinewas prepared in two steps by reaction of trifluoromethylcopper with 3-chloro-4-iodonitrobenzene.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41252-96-4
Molecular Formula C6H3ClINO2
Molecular Weight (g/mol) 283.449
MDL Number MFCD00024589
InChI Key FQXFHRSYMORXKN-UHFFFAOYSA-N
Synonym 3-chloro-4-iodonitrobenzene, 3-chloro-4-iodo-nitrobenzene, 3-chloro-4-iodo-nitro-benzene, benzene, 2-chloro-1-iodo-4-nitro, 2-chlor-1-iod-4-nitrobenzol, acmc-20a36k, 3-chloro-4-iodonitro benzene, 1-iodo-2-chloro-4-nitrobenzene, 2-chloro-1-iodo-4-nitro-benzene, 1-chloro-2-iodo-5-nitrobenzene
PubChem CID 96640
IUPAC Name 2-chloro-1-iodo-4-nitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)I

Specifications

Melting Point 97°C to 101°C
Quantity 1 g
Beilstein 2502411
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 283.45
Percent Purity 98%
Chemical Name or Material 2-Chloro-1-iodo-4-nitrobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.