missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-fluoro-4-iodobenzene, 98%

Product Code. 11419403
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419403 25 g 25g
11429403 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419403 Supplier Thermo Scientific Alfa Aesar Supplier No. A11808.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 156150-67-3 | C6H3ClFI | 256.443 g/mol

It employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156150-67-3
Molecular Formula C6H3ClFI
Molecular Weight (g/mol) 256.443
MDL Number MFCD00042210
InChI Key OMASDGWBVAVFQZ-UHFFFAOYSA-N
Synonym 3-chloro-4-fluoroiodobenzene, 1-chloro-2-fluoro-5-iodobenzene, 2-chloro4-iodofluorobenzene, 3-chloro-4-fluoro-1-iodobenzene, 1-fluoro-2-chloro-4-iodobenzene, benzene, 2-chloro-1-fluoro-4-iodo, benzene,2-chloro-1-fluoro-4-iodo, 2-chloro-1-fluoro-4-iodo-benzene, pubchem3433, acmc-20an6s
PubChem CID 2724605
IUPAC Name 2-chloro-1-fluoro-4-iodobenzene
SMILES C1=CC(=C(C=C1I)Cl)F

Specifications

Density 2.02
Boiling Point 94°C to 95°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.605
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 256.45
Percent Purity 98%
Chemical Name or Material 2-Chloro-1-fluoro-4-iodobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.