missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-thiazole-5-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10758055
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10758055

Brand: Thermo Scientific Maybridge CC62902DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 148637-74-5
Molecular Formula C4HCl2NOS
Molecular Weight (g/mol) 182.018
MDL Number MFCD09879938
InChI Key GZSJPOBGZMUSPV-UHFFFAOYSA-N
Synonym 5-thiazolecarbonylchloride, 2-chloro, acmc-20n5hc, 2-chlorothiazole-5-carbonylchloride, 2-chlorothiazole-5-carbonyl chloride, 2-chloro-thiazole-5-carbonyl chloride, 5-thiazolecarbonyl chloride, 2-chloro, 5-thiazolecarbonyl chloride, 2-chloro-9ci
PubChem CID 24229701
IUPAC Name 2-chloro-1,3-thiazole-5-carbonyl chloride
SMILES C1=C(SC(=N1)Cl)C(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 182.03
Percent Purity 97%
Chemical Name or Material 2-Chloro-1, 3-thiazole-5-carbonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.